Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205241.1 | NP_001192170.1 | 3 - 4 | 1031 | 71 | |
NM_017479.3 | NP_059507.2 | 3 - 4 | 1155 | 71 | ||
GenBank mRNA | AF222800.1 | - | 3 - 4 | 1071 | 71 | |
AK045188.1 | - | 3 - 4 | 1167 | 71 | ||
AK083123.1 | - | 3 - 4 | 1170 | 71 | ||
AK135465.1 | - | 3 - 4 | 1207 | 71 | ||
AK147509.1 | - | 3 - 4 | 1032 | 71 | ||
BC095974.1 | - | 2 - 3 | 808 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |