Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019786.4 | NP_062760.3 | 15 - 16 | 1873 | 138 | |
GenBank mRNA | AF145705.1 | - | 14 - 15 | 1725 | 138 | |
AF191839.1 | - | 14 - 15 | 1752 | 138 | ||
AK004649.2 | - | 15 - 16 | 1827 | 138 | ||
AK083214.1 | - | 15 - 16 | 1821 | 138 | ||
AK168193.1 | - | 15 - 16 | 1855 | 138 | ||
BC129910.1 | - | 14 - 15 | 1785 | 40111752 | 138 | |
BC129911.1 | - | 14 - 15 | 1785 | 40111754 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |