Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmfbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019938.3 | NP_064322.1 | 11 - 12 | 1768 | 89 | |
GenBank mRNA | AB029919.1 | - | 11 - 12 | 1768 | 89 | |
AK006021.1 | - | 2 - 3 | 219 | 89 | ||
BC138245.1 | - | 11 - 12 | 1720 | 8861264 | 89 | |
BC138246.1 | - | 11 - 12 | 1720 | 8861265 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |