Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019953.1 | NP_064337.1 | 2 - 3 | 433 | 73 | |
GenBank mRNA | AF186115.1 | - | 2 - 3 | 433 | 73 | |
AK007914.1 | - | 2 - 3 | 194 | 73 | ||
AK013014.1 | - | 2 - 3 | 304 | 73 | ||
AK013568.1 | - | 2 - 3 | 175 | 73 | ||
AK019927.1 | - | 2 - 3 | 305 | 73 | ||
AK150953.1 | - | 2 - 3 | 377 | 73 | ||
BC008261.1 | - | 2 - 3 | 205 | 2650612 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |