Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128080.2 | NP_001121552.1 | 4 - 5 | 414 | 69 | |
NM_001128081.1 | NP_001121553.1 | 3 - 4 | 529 | 69 | ||
NM_020286.3 | NP_064682.1 | 3 - 4 | 529 | 69 | ||
GenBank mRNA | AF175771.1 | - | 3 - 4 | 378 | 69 | |
AF291425.1 | - | 3 - 4 | 497 | 69 | ||
AJ279791.1 | - | 3 - 4 | 489 | 69 | ||
AJ279792.1 | - | 3 - 4 | 489 | 69 | ||
AJ279793.1 | - | 3 - 4 | 489 | 69 | ||
AK013248.1 | - | 3 - 4 | 305 | 69 | ||
AK154694.1 | - | 3 - 4 | 550 | 69 | ||
AY039000.1 | - | 4 - 5 | 422 | 69 | ||
BC055858.1 | - | 3 - 4 | 503 | 4913671 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |