Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021607.3 | NP_067620.3 | 4 - 5 | 498 | 74 | |
GenBank mRNA | AF240469.1 | - | 4 - 5 | 448 | 74 | |
AK033314.1 | - | 4 - 5 | 440 | 74 | ||
AK129102.1 | - | 4 - 5 | 498 | 74 | ||
AK170173.1 | - | 4 - 5 | 444 | 74 | ||
AK170244.1 | - | 4 - 5 | 463 | 74 | ||
AK170369.1 | - | 4 - 5 | 487 | 74 | ||
AK172579.1 | - | 4 - 5 | 443 | 74 | ||
BC019998.1 | - | 4 - 5 | 443 | 4458780 | 74 | |
CD549803.1 | - | 4 - 5 | 498 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |