Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021793.2 | NP_068565.1 | 10 - 11 | 1890 | 93 | |
GenBank mRNA | AB045293.1 | - | 10 - 11 | 1889 | 93 | |
AK044384.1 | - | 10 - 11 | 1891 | 93 | ||
BC005722.1 | - | 9 - 10 | 1766 | 3595205 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |