Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289575.1 | NP_001276504.1 | 21 - 22 | 3040 | 118 | |
NM_001289576.1 | NP_001276505.1 | 21 - 22 | 3022 | 118 | ||
NM_022887.4 | NP_075025.2 | 21 - 22 | 3008 | 118 | ||
GenBank mRNA | AB047561.1 | - | 19 - 20 | 2807 | 118 | |
AJ271912.1 | - | 21 - 22 | 3032 | 118 | ||
AK030318.1 | - | 21 - 22 | 3005 | 118 | ||
AK122229.1 | - | 7 - 8 | 1332 | 118 | ||
AK147428.1 | - | 21 - 22 | 3013 | 118 | ||
BC052399.1 | - | 20 - 21 | 2961 | 5720967 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |