Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023718.3 | NP_076207.1 | 8 - 9 | 1074 | 102 | |
GenBank mRNA | AF203701.1 | - | 8 - 9 | 1078 | 102 | |
AF213260.1 | - | 8 - 9 | 1078 | 102 | ||
AK142587.1 | - | 7 - 8 | 932 | 102 | ||
BC071214.1 | - | 7 - 8 | 924 | 5042082 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |