Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppil4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026141.3 | NP_080417.2 | 7 - 8 | 738 | 128 | |
GenBank mRNA | AK011443.1 | - | 7 - 8 | 710 | 128 | |
AK012371.1 | - | 7 - 8 | 715 | 128 | ||
AK014406.2 | - | 711 | 128 | |||
AK014453.1 | - | 6 - 7 | 1263 | 128 | ||
AK017370.1 | - | 7 - 8 | 738 | 128 | ||
AK167168.1 | - | 7 - 8 | 703 | 128 | ||
AK167258.1 | - | 7 - 8 | 710 | 128 | ||
BC079912.1 | - | 7 - 8 | 700 | 30661438 | 128 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |