Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekha3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031256.3 | NP_112546.1 | 5 - 6 | 901 | 67 | |
GenBank mRNA | AF286163.1 | - | 5 - 6 | 690 | 67 | |
BC012283.1 | - | 4 - 5 | 1160 | 67 | ||
BC031110.1 | - | 6 - 7 | 709 | 4981486 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |