Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001256382.1 | NP_001243311.1 | 15 - 16 | 3101 | 88 | |
NM_001256383.1 | NP_001243312.1 | 15 - 16 | 3101 | 88 | ||
NM_053271.2 | NP_444501.1 | 17 - 18 | 3221 | 88 | ||
GenBank mRNA | AB021131.1 | - | 17 - 18 | 3175 | 88 | |
AK129206.1 | - | 12 - 13 | 2187 | 88 | ||
BC118979.2 | - | 15 - 16 | 3049 | 40098740 | 88 | |
HM015529.1 | - | 15 - 16 | 3024 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |