Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Recql4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058214.3 | NP_478121.2 | 19 - 20 | 3349 | 69 | |
GenBank mRNA | AB039882.1 | - | 18 - 19 | 3375 | 69 | |
AB175741.1 | - | 19 - 20 | 3266 | 69 | ||
AK090260.1 | - | 19 - 20 | 3031 | 69 | ||
BC064680.1 | - | 19 - 20 | 3323 | 69 | ||
BC131656.1 | - | 19 - 20 | 3262 | 69 | ||
BC131657.1 | - | 17 - 18 | 3599 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |