Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nobox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130869.3 | NP_570939.1 | 6 - 7 | 763 | 70 | |
GenBank mRNA | AB221582.1 | - | 6 - 7 | 740 | 70 | |
AY061761.1 | - | 6 - 7 | 771 | 70 | ||
BC125280.1 | - | 6 - 7 | 759 | 40129795 | 70 | |
BC125282.1 | - | 6 - 7 | 759 | 40129797 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |