Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mlc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133241.2 | NP_573504.1 | 2 - 3 | 233 | 65 | |
GenBank mRNA | AF449425.1 | - | 2 - 3 | 687 | 65 | |
AK030396.1 | - | 2 - 3 | 232 | 65 | ||
AK043757.1 | - | 2 - 3 | 233 | 65 | ||
AK122190.1 | - | 2 - 3 | 675 | 65 | ||
BC024719.1 | - | 2 - 3 | 233 | 5363238 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |