Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133254.3 | NP_573517.1 | 5 - 6 | 566 | 81 | |
GenBank mRNA | AK128900.1 | - | 5 - 6 | 580 | 81 | |
AY033886.1 | - | 5 - 6 | 576 | 81 | ||
BC022226.1 | - | 5 - 6 | 566 | 4235707 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |