Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shkbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138676.2 | NP_619617.2 | 6 - 7 | 455 | 64 | |
GenBank mRNA | AF246218.1 | - | 6 - 7 | 461 | 64 | |
AK047434.1 | - | 4 - 5 | 528 | 64 | ||
AK154779.1 | - | 6 - 7 | 444 | 64 | ||
AK160641.1 | - | 5 - 6 | 527 | 64 | ||
BC061477.1 | - | 6 - 7 | 408 | 6336438 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |