Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145138.2 | NP_660120.2 | 19 - 20 | 2604 | 88 | |
GenBank mRNA | AJ489828.1 | - | 19 - 20 | 2579 | 88 | |
AK173323.1 | - | 19 - 20 | 2588 | 88 | ||
BC024926.1 | - | 1 - 2 | 191 | 88 | ||
BC117971.2 | - | 19 - 20 | 2545 | 40098375 | 88 | |
BC117972.2 | - | 19 - 20 | 2544 | 40098378 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |