Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145630.2 | NP_663605.1 | 2 - 3 | 395 | 84 | |
GenBank mRNA | AK075879.1 | - | 2 - 3 | 391 | 84 | |
AK075985.1 | - | 2 - 3 | 451 | 84 | ||
AK088478.1 | - | 2 - 3 | 476 | 84 | ||
AK132024.1 | - | 2 - 3 | 419 | 84 | ||
AK169874.1 | - | 2 - 3 | 476 | 84 | ||
BC008126.1 | - | 2 - 3 | 395 | 3500763 | 84 | |
CT010338.1 | - | 2 - 3 | 252 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |