Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp9y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148943.2 | NP_683745.2 | 33 - 34 | 5248 | 123 | |
GenBank mRNA | AJ307017.1 | - | 33 - 34 | 5248 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mus musculus Probe spans exons |