Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cited1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276466.1 | NP_001263395.1 | 1 - 2 | 149 | 86 | |
GenBank mRNA | AK148042.1 | - | 1 - 2 | 131 | 86 | |
U65091.1 | - | 1 - 2 | 131 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |