Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008230.6 | NP_032256.3 | 10 - 11 | 1270 | 99 | |
GenBank mRNA | AF109137.1 | - | 10 - 11 | 1180 | 99 | |
AK088545.1 | - | 10 - 11 | 1257 | 99 | ||
AK133455.1 | - | 10 - 11 | 1271 | 99 | ||
AK150168.1 | - | 10 - 11 | 1258 | 99 | ||
AK153104.1 | - | 10 - 11 | 1258 | 99 | ||
BC052833.1 | - | 10 - 11 | 1255 | 30044098 | 99 | |
X57437.1 | - | 10 - 11 | 1235 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |