Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB015290.1 | - | 3 - 4 | 317 | 61 | |
AF068182.1 | - | 3 - 4 | 298 | 61 | ||
AJ222814.1 | - | 3 - 4 | 268 | 61 | ||
AK089704.1 | - | 3 - 4 | 283 | 61 | ||
BC059785.1 | - | 3 - 4 | 269 | 5339723 | 61 | |
Y17159.1 | - | 3 - 4 | 643 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |