Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008576.3 | NP_032602.1 | 15 - 16 | 2025 | 90 | |
GenBank mRNA | AF022908.1 | - | 15 - 16 | 1987 | 90 | |
AK029876.1 | - | 15 - 16 | 2026 | 90 | ||
BC090617.1 | - | 15 - 16 | 1996 | 30943198 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |