Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008576.3 | NP_032602.1 | 21 - 22 | 2898 | 63 | |
GenBank mRNA | AF022908.1 | - | 21 - 22 | 2860 | 63 | |
AK029876.1 | - | 21 - 22 | 2899 | 63 | ||
AK049174.1 | - | 5 - 6 | 665 | 63 | ||
BC090617.1 | - | 21 - 22 | 2869 | 30943198 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |