Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009202.5 | NP_033228.2 | 7 - 8 | 1456 | 62 | |
GenBank mRNA | AF010259.1 | - | 7 - 8 | 1314 | 62 | |
BC021651.1 | - | 7 - 8 | 1456 | 4211979 | 62 | |
U38652.1 | - | 7 - 8 | 1311 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |