Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp61 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009561.2 | NP_033587.2 | 5 - 6 | 802 | 105 | |
GenBank mRNA | AK148158.1 | - | 5 - 6 | 105 | ||
AK149960.1 | - | 5 - 6 | 597 | 105 | ||
BC006618.1 | - | 5 - 6 | 841 | 3967527 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |