Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009619.4 | NP_033749.2 | 13 - 14 | 1414 | 91 | |
GenBank mRNA | BC129920.1 | - | 13 - 14 | 1380 | 91 | |
KF144395.1 | - | 2 - 3 | 163 | 91 | ||
KF144396.1 | - | 2 - 3 | 163 | 91 | ||
X64227.1 | - | 13 - 14 | 1424 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |