Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010401.3 | NP_034531.1 | 16 - 17 | 1882 | 76 | |
GenBank mRNA | AK014518.1 | - | 16 - 17 | 1882 | 76 | |
AK028963.1 | - | 16 - 17 | 1589 | 76 | ||
BC057637.1 | - | 16 - 17 | 1559 | 4211545 | 76 | |
BC157999.1 | - | 16 - 17 | 1864 | 9088147 | 76 | |
L07645.1 | - | 16 - 17 | 1566 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |