Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zpr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011752.2 | NP_035882.1 | 5 - 6 | 714 | 71 | |
GenBank mRNA | AB041612.1 | - | 5 - 6 | 597 | 71 | |
AK044472.1 | - | 5 - 6 | 603 | 71 | ||
AK158995.1 | - | 5 - 6 | 675 | 71 | ||
AK167849.1 | - | 5 - 6 | 585 | 71 | ||
AK218200.1 | - | 3 - 4 | 189 | 71 | ||
BC021397.1 | - | 5 - 6 | 597 | 5067274 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |