Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291025.1 | NP_001277954.1 | 10 - 11 | 1892 | 73 | |
NM_011822.4 | NP_035952.2 | 9 - 10 | 1712 | 73 | ||
GenBank mRNA | AB008895.1 | - | 9 - 10 | 1667 | 73 | |
AK155173.1 | - | 8 - 9 | 1893 | 73 | ||
AK163680.1 | - | 9 - 10 | 1688 | 73 | ||
AK169284.1 | - | 9 - 10 | 1695 | 73 | ||
AK170431.1 | - | 10 - 11 | 1839 | 73 | ||
AK218271.1 | - | 1 - 2 | 109 | 73 | ||
BC003917.1 | - | 7 - 8 | 802 | 73 | ||
BC014287.1 | - | 9 - 10 | 1685 | 4193343 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |