Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Unc93b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161428.1 | NP_001154900.1 | 7 - 8 | 978 | 57 | |
NM_019449.2 | NP_062322.2 | 7 - 8 | 978 | 57 | ||
GenBank mRNA | AJ422144.1 | - | 1 - 2 | 120 | 57 | |
AK030488.1 | - | 7 - 8 | 939 | 57 | ||
AK155556.1 | - | 7 - 8 | 936 | 57 | ||
AK162535.1 | - | 7 - 8 | 931 | 57 | ||
AK165060.1 | - | 7 - 8 | 938 | 57 | ||
BC018388.1 | - | 7 - 8 | 945 | 4209296 | 57 | |
BC023731.1 | - | 1 - 2 | 403 | 5352340 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |