Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290448.1 | NP_001277377.1 | 3 - 4 | 1304 | 57 | |
NM_001290449.1 | NP_001277378.1 | 3 - 4 | 1304 | 57 | ||
NM_023739.3 | NP_076228.2 | 3 - 4 | 1267 | 57 | ||
GenBank mRNA | AF223576.1 | - | 3 - 4 | 1231 | 57 | |
AK005038.2 | - | 3 - 4 | 1239 | 57 | ||
AK033850.1 | - | 3 - 4 | 1198 | 57 | ||
AK050749.1 | - | 3 - 4 | 1241 | 57 | ||
AK144647.1 | - | 3 - 4 | 1253 | 57 | ||
AK146108.1 | - | 3 - 4 | 1221 | 57 | ||
AK155387.1 | - | 3 - 4 | 1219 | 57 | ||
AK160013.1 | - | 3 - 4 | 1237 | 57 | ||
BC053709.1 | - | 3 - 4 | 1202 | 30052746 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |