Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025532.3 | NP_079808.1 | 1 - 2 | 96 | 61 | |
GenBank mRNA | AF525155.1 | - | 1 - 2 | 47 | 61 | |
AK005411.1 | - | 1 - 2 | 97 | 61 | ||
AK009863.1 | - | 1 - 2 | 97 | 61 | ||
BC100329.1 | - | 1 - 2 | 86 | 30787440 | 61 | |
BC111112.1 | - | 1 - 2 | 92 | 40050224 | 61 | |
BC138199.1 | - | 1 - 2 | 89 | 8861218 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |