Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026390.2 | NP_080666.2 | 11 - 12 | 1473 | 93 | |
GenBank mRNA | AK032206.1 | - | 11 - 12 | 1351 | 93 | |
AK075716.1 | - | 11 - 12 | 1208 | 93 | ||
AK129098.1 | - | 2 - 3 | 247 | 93 | ||
AK145551.1 | - | 8 - 9 | 899 | 93 | ||
BC019795.1 | - | 11 - 12 | 1492 | 4158848 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |