Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifih1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164477.1 | NP_001157949.1 | 12 - 13 | 2795 | 99 | |
NM_027835.3 | NP_082111.2 | 13 - 14 | 2942 | 99 | ||
GenBank mRNA | AF374384.1 | - | 13 - 14 | 2851 | 99 | |
AK153018.1 | - | 13 - 14 | 2849 | 99 | ||
AK153344.1 | - | 5 - 6 | 876 | 99 | ||
AY075132.1 | - | 13 - 14 | 2617 | 99 | ||
BC004031.1 | - | 6 - 7 | 1084 | 99 | ||
BC025508.1 | - | 8 - 9 | 1386 | 99 | ||
BC080200.1 | - | 12 - 13 | 2700 | 3597815 | 99 | |
EF113547.1 | - | 13 - 14 | 2631 | 99 | ||
EF113548.1 | - | 13 - 14 | 2631 | 99 | ||
EU084750.1 | - | 13 - 14 | 2631 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |