Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053253.3 | NP_444483.2 | 4 - 5 | 522 | 120 | |
GenBank mRNA | AF123386.1 | - | 2 - 3 | 153 | 120 | |
AK132332.1 | - | 4 - 5 | 531 | 120 | ||
BC052357.1 | - | 4 - 5 | 522 | 6772144 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |