Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133345.2 | NP_579923.1 | 1 - 2 | 54 | 84 | |
GenBank mRNA | AK002821.1 | - | 1 - 2 | 56 | 84 | |
AK009267.1 | - | 1 - 2 | 55 | 84 | ||
AK031633.1 | - | 1 - 2 | 59 | 84 | ||
AK046126.1 | - | 1 - 2 | 67 | 84 | ||
AK050522.1 | - | 1 - 2 | 67 | 84 | ||
AY035880.1 | - | 1 - 2 | 41 | 84 | ||
AY035881.1 | - | 1 - 2 | 41 | 84 | ||
AY036107.1 | - | 1 - 2 | 41 | 84 | ||
BC009127.1 | - | 1 - 2 | 42 | 3708722 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |