Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145223.2 | NP_660258.2 | 8 - 9 | 5125 | 107 | |
GenBank mRNA | AF425257.1 | - | 8 - 9 | 5126 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |