Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnpla7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146251.4 | NP_666363.3 | 33 - 34 | 4083 | 64 | |
GenBank mRNA | AK088362.1 | - | 33 - 34 | 4029 | 64 | |
AK148380.1 | - | 33 - 34 | 4057 | 64 | ||
AK170417.1 | - | 33 - 34 | 4029 | 64 | ||
BC021312.1 | - | 32 - 33 | 4117 | 64 | ||
BC025621.1 | - | 29 - 30 | 3496 | 64 | ||
BC027342.1 | - | 11 - 12 | 1527 | 64 | ||
BC064003.1 | - | 16 - 17 | 1847 | 64 | ||
BC080793.1 | - | 11 - 12 | 1370 | 64 | ||
BC141365.1 | - | 33 - 34 | 3867 | 9055978 | 64 | |
BC141366.1 | - | 33 - 34 | 3867 | 9055979 | 64 | |
EF685370.1 | - | 9 - 10 | 988 | 64 | ||
EU046569.1 | - | 13 - 14 | 1644 | 64 | ||
EU293208.1 | - | 32 - 33 | 3754 | 64 | ||
EU368675.1 | - | 32 - 33 | 3754 | 64 | ||
EU402948.2 | - | 31 - 32 | 3690 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |