Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148951.1 | NP_683753.1 | 2 - 3 | 376 | 75 | |
GenBank mRNA | AB074494.1 | - | 2 - 3 | 376 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |