Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153128.2 | NP_694768.1 | 2 - 3 | 351 | 85 | |
GenBank mRNA | AK034170.1 | - | 2 - 3 | 339 | 85 | |
AK165280.1 | - | 2 - 3 | 329 | 85 | ||
BC034514.1 | - | 2 - 3 | 351 | 3157064 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |