Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153128.2 | NP_694768.1 | 8 - 9 | 1289 | 64 | |
GenBank mRNA | AK034170.1 | - | 8 - 9 | 1277 | 64 | |
BC034514.1 | - | 8 - 9 | 1289 | 3157064 | 64 | |
BC038811.1 | - | 2 - 3 | 317 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |