Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153172.1 | NP_694812.1 | 2 - 3 | 178 | 84 | |
GenBank mRNA | AF392062.1 | - | 2 - 3 | 178 | 84 | |
AK077121.1 | - | 2 - 3 | 176 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Recommended Assay Mus musculus Probe spans exons |