Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153391.2 | NP_700440.2 | 23 - 24 | 2703 | 71 | |
GenBank mRNA | AK052303.1 | - | 23 - 24 | 2683 | 71 | |
AK129413.1 | - | 15 - 16 | 1792 | 71 | ||
AK147970.1 | - | 23 - 24 | 2703 | 71 | ||
AY029258.1 | - | 23 - 24 | 2773 | 71 | ||
BC046432.1 | - | 8 - 9 | 905 | 71 | ||
BC060134.1 | - | 23 - 24 | 2697 | 71 | ||
BC082301.1 | - | 22 - 23 | 2583 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |