Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163024.1 | NP_001156496.1 | 6 - 7 | 1202 | 62 | |
NM_001163025.1 | NP_001156497.1 | 6 - 7 | 1202 | 62 | ||
NM_153395.2 | NP_700444.2 | 6 - 7 | 1202 | 62 | ||
GenBank mRNA | AB014059.1 | - | 6 - 7 | 724 | 62 | |
AK122425.1 | - | 6 - 7 | 1202 | 62 | ||
AK161217.1 | - | 6 - 7 | 1081 | 62 | ||
BC064131.1 | - | 6 - 7 | 692 | 62 | ||
BC150943.1 | - | 6 - 7 | 723 | 9087854 | 62 | |
CF747261.1 | - | 2 - 3 | 133 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |