Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Specc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145826.1 | NP_001139298.1 | 9 - 10 | 2636 | 83 | |
NM_153406.3 | NP_700455.3 | 6 - 7 | 2407 | 83 | ||
GenBank mRNA | AB093236.1 | - | 9 - 10 | 2636 | 83 | |
AK041543.1 | - | 3 - 4 | 900 | 83 | ||
AY884297.1 | - | 6 - 7 | 2408 | 83 | ||
BC037464.1 | - | 3 - 4 | 1494 | 4194868 | 83 | |
BC072595.1 | - | 5 - 6 | 2246 | 6408832 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |