Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153781.1 | NP_722476.1 | 1 - 2 | 316 | 68 | |
GenBank mRNA | AK032769.1 | - | 1 - 2 | 329 | 68 | |
AK035673.1 | - | 1 - 2 | 45 | 68 | ||
AK134667.1 | - | 1 - 2 | 326 | 68 | ||
AK142716.1 | - | 1 - 2 | 325 | 68 | ||
AK147763.1 | - | 1 - 2 | 348 | 68 | ||
AK168381.1 | - | 1 - 2 | 326 | 68 | ||
AK169052.1 | - | 1 - 2 | 68 | |||
AK169631.1 | - | 1 - 2 | 330 | 68 | ||
BC031457.1 | - | 1 - 2 | 324 | 68 | ||
BC035283.1 | - | 1 - 2 | 316 | 4947455 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |