Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153795.2 | NP_722490.1 | 5 - 6 | 895 | 79 | |
GenBank mRNA | AK090332.1 | - | 5 - 6 | 792 | 79 | |
AK142912.1 | - | 5 - 6 | 896 | 79 | ||
AK149737.1 | - | 5 - 6 | 826 | 79 | ||
AK158357.1 | - | 5 - 6 | 791 | 79 | ||
AK169682.1 | - | 5 - 6 | 806 | 79 | ||
BC025119.1 | - | 5 - 6 | 827 | 4190760 | 79 | |
BC032204.1 | - | 5 - 6 | 871 | 5342463 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |