Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153803.1 | NP_722498.1 | 4 - 5 | 445 | 65 | |
GenBank mRNA | AK136711.1 | - | 4 - 5 | 479 | 65 | |
AK143057.1 | - | 3 - 4 | 376 | 65 | ||
BC038479.1 | - | 4 - 5 | 445 | 4503030 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |